1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Carboxy-1-methyl-3-phenyl-1H-pyrazole

A155876-100MG A155876-250MG A155876-1G A155876-5G
103804-810EA 48 USD
103804-810 103843-012 77235-640 77235-642 77318-752 77318-754
1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid 97%
1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.209 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 186 °C
Flash Pt: 207.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08271939
CAS Number: 10250-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR