1-Methyl-3,4-dihydroisoquinoline 95%

Supplier: Ambeed
Warning

A101415-1G A101415-250MG
103795-964EA 64 USD
103795-964 103821-648 77297-098 77891-750
1-Methyl-3,4-dihydroisoquinoline 95%
1-Methyl-3,4-dihydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₁N
Boiling Pt: 229.8 °C (760 mm Hg)
Flash Pt: 84.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00047648
CAS Number: 2412-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR