2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid

A100707-100MG A100707-1G A100707-5G A100707-250MG
103795-584EA 54 USD
103795-584 103821-282 77270-168 77296-820 77296-822 77747-752
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 97%
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂O₂
MW: 194.11 g/mol
Boiling Pt: 288.8 °C (760 mm Hg)
Melting Pt: 130…132 °C
Flash Pt: 128.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02090853
CAS Number: 128694-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR