1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-3-(trifluoromethyl)pyrazole-4-carboxylic acid
A112969-100G
A112969-10G
A112969-250MG
A112969-25G
A112969-1G
A112969-5G
103798-140EA
6
USD
103798-140
103823-974
77233-676
77233-678
77233-680
77233-682
77310-808
77310-810
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 98%
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₅F₃N₂O₂ MW: 194.1113 g/mol Boiling Pt: 285.6 °C (760 mm Hg) Melting Pt: 203 °C Flash Pt: 126.51 °C Storage Temperature: Ambient |
MDL Number:
MFCD01936005 CAS Number: 113100-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...