1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrazole-3-boronic acid pinacol ester

A117827-100MG A117827-250MG A117827-1G A117827-5G A117827-10G A117827-25G
103798-940EA 14 USD
103798-940 103824-832 76971-330 76971-332 77311-610 77311-612 77566-152 77670-660
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: For research use only.

Formula: C₁₀H₁₇BN₂O₂
Boiling Pt: 308.3 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04114000
CAS Number: 1020174-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR