1-Methylpyrrole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-pyrrole-2-carboxylic acid, 1-Methyl-2-pyrrolecarboxylic acid

A123267-10G A123267-250MG A123267-25G A123267-1G A123267-5G
103800-376EA 8 USD
103800-376 76810-642 76810-644 76810-646 77312-494 77312-496
1-Methylpyrrole-2-carboxylic acid 98%
1-Methylpyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Boiling Pt: 276.6 °C (760 mm Hg)
Melting Pt: 131…138 °C
Flash Pt: 121.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00003088
CAS Number: 6973-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR