1-Methyl-2-oxopyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A927164-10G A927164-1G A927164-250MG
103841-412EA 58 USD
103841-412 76880-892 77308-522 77674-384
1-Methyl-2-oxopyrrolidine-3-carboxylic acid 97%
1-Methyl-2-oxopyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.141 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19228580
CAS Number: 30932-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR