5-Amino-1-methyltetrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1-Methyl-1H-tetrazol-5-amine

A659235-1G A659235-5G A659235-10G
103840-432EA 13 USD
103840-432 76844-624 76844-626 77749-356
5-Amino-1-methyltetrazole 98%
5-Amino-1-methyltetrazole

Precaución: for Research Use Only

Formula: C₂H₅N₅
MW: 99.10 g/mol
Boiling Pt: 282.9 °C (760 mm Hg)
Melting Pt: 226…230 °C
Flash Pt: 124.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00186454
CAS Number: 5422-44-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR