1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A304720-1G A304720-5G A304720-10G A304720-250MG
103814-420EA 23 USD
103814-420 103835-590 76838-286 76838-288 77239-310 77239-312
1-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
1-Methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Boiling Pt: 254.2 °C (760 mm Hg)
Flash Pt: 107.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09743451
CAS Number: 27257-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR