N-Methylmaleimide 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-pyrrole-2,5-dione

A266942-100G A266942-10G A266942-25G A266942-1G A266942-5G
103812-822EA 6 USD
103812-822 103852-724 76722-960 76722-962 77092-438 77304-962 77304-964
N-Methylmaleimide 97%
N-Methylmaleimide

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MW: 111.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005508
CAS Number: 930-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR