1-Methyl-1H-pyrazole-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A120274-100G
A120274-10G
A120274-25G
A120274-1G
A120274-5G
103799-906EA
8
USD
103799-906
76983-210
76983-212
76983-214
77312-010
77312-012
1-Methyl-1H-pyrazole-5-carboxylic acid 98%
1-Methyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₆N₂O₂ Melting Pt: 223.5…224.5 °C Flash Pt: 139.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464253 CAS Number: 16034-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...