1-Methyl-1H-pyrazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrazole-5-carboxaldehyde, 5-Formyl-1-methylpyrazole

A462986-1G A462986-25G A462986-5G A462986-100G A462986-10G
103827-946EA 13 USD
103827-946 103839-022 76842-912 76842-914 76842-916 77168-990 77168-992
1-Methyl-1H-pyrazole-5-carbaldehyde 97%
1-Methyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.1100 g/mol
Boiling Pt: 218.9 °C (760 mm Hg)
Flash Pt: 86.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03419801
CAS Number: 27258-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR