1-Methyl-1H-pyrazole-4-sulfonyl chloride 98%

Supplier: Ambeed
Danger

A103074-10G A103074-250MG A103074-25G A103074-1G A103074-5G
103796-478EA 27 USD
103796-478 103822-180 77025-236 77025-238 77025-240 77309-050 77309-052
1-Methyl-1H-pyrazole-4-sulfonyl chloride 98%
1-Methyl-1H-pyrazole-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O₂S
MW: 179.98 g/mol
Flash Pt: 174.066 °C
Storage Temperature: Ambient
MDL Number: MFCD04968667
CAS Number: 288148-34-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR