1-Methyl-1H-pyrazol-4-carbaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-1H-pyrazole-4-carbaldehyde, 1-Methyl-1H-pyrazole-4-carboxaldehyde
A369202-25G
A369202-5G
A369202-100G
A369202-10G
A369202-1G
103826-448EA
5
USD
103826-448
103837-462
76840-710
76840-712
77167-940
77167-942
77306-802
1-Methyl-1H-pyrazol-4-carbaldehyde 98%
1-Methyl-1H-pyrazol-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₅H₆N₂O MW: 110.12 g/mol Melting Pt: 160…165 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00460465 CAS Number: 25016-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...