1-Methyl-1H-pyrazole-3-sulfonyl chloride 97%

Supplier: Ambeed
Danger

A156152-100MG A156152-10G A156152-250MG A156152-1G A156152-25G A156152-5G
103804-838EA 44 USD
103804-838 103843-042 77235-672 77235-674 77235-676 77318-790 77318-792 77318-794
1-Methyl-1H-pyrazole-3-sulfonyl chloride 97%
1-Methyl-1H-pyrazole-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O₂S
MW: 180.6127 g/mol
Boiling Pt: 295.6 °C (760 mm Hg)
Flash Pt: 132.618 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690273
CAS Number: 89501-90-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR