1-Methyl-1H-pyrazole-3-carbaldehyde 98%
Supplier: Ambeed
|
Warning
|
A389026-1G
A389026-25G
A389026-100G
A389026-10G
103826-844EA
9
USD
103826-844
103837-884
76841-246
76841-248
77168-166
77168-168
1-Methyl-1H-pyrazole-3-carbaldehyde 98%
1-Methyl-1H-pyrazole-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₅H₆N₂O MW: 110.1100 g/mol Boiling Pt: 280.3 °C (760 mm Hg) Flash Pt: 86.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03419800 CAS Number: 27258-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...