1-Methyl-1H-pyrazol-5-ol 97%

Supplier: Ambeed
Warning

A151228-100G A151228-10G A151228-250MG A151228-500G A151228-1G A151228-25G A151228-5G
103804-238EA 6 USD
103804-238 103842-386 76768-158 76768-160 76768-162 76768-164 77317-924 77317-926 77317-928
1-Methyl-1H-pyrazol-5-ol 97%
1-Methyl-1H-pyrazol-5-ol

Precaución: for Research Use Only

Formula: C₄H₆N₂O
Boiling Pt: 217.6 °C (760 mm Hg)
Melting Pt: 110…114 °C
Flash Pt: 85.439 °C
Storage Temperature: Ambient
MDL Number: MFCD05864418
CAS Number: 33641-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR