4-Hydroxy-1-methyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-pyrazol-4-ol

A170574-1G A170574-100MG A170574-250MG
103806-516EA 30 USD
103806-516 103844-790 77081-902 77885-339 77885-340
4-Hydroxy-1-methyl-1H-pyrazole 95%
4-Hydroxy-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₆N₂O
Boiling Pt: 217.7±13.0 °C (760 mm Hg)
Flash Pt: 85.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22414500
CAS Number: 78242-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR