4-Amino-1-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-methyl-1H-pyrazol-4-amine

A152410-10G A152410-1G A152410-25G A152410-5G
103804-390EA 6 USD
103804-390 103842-558 76783-428 77299-948 77318-170 77318-172
4-Amino-1-methyl-1H-pyrazole 98%
4-Amino-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 231.4 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD05667135
CAS Number: 69843-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR