3-Amino-1-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A161773-100G A161773-10G A161773-1G A161773-25G A161773-5G
103805-536EA 5 USD
103805-536 103843-778 76997-826 76997-828 77319-786 77319-788 77319-790
3-Amino-1-methyl-1H-pyrazole 98%
3-Amino-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₇N₃
Boiling Pt: 231.4 °C (760 mm Hg)
Flash Pt: 93.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00466340
CAS Number: 1904-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR