1-Methyl-1H-indole-6-carbaldehyde 95%

Supplier: Ambeed
Warning

A275454-250MG A275454-5G
103813-254EA 59 USD
103813-254 103853-180 77032-028 77305-194
1-Methyl-1H-indole-6-carbaldehyde 95%
1-Methyl-1H-indole-6-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690251
CAS Number: 21005-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR