1-methyl-1H-indole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A698121-5G A698121-100MG A698121-250MG
103840-616EA 187 USD
103840-616 76844-840 77035-758 77035-760
1-methyl-1H-indole-5-carbaldehyde 97%
1-methyl-1H-indole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.188 g/mol
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 84…85.5 °C at N/A mmHg
Flash Pt: 146.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08271908
CAS Number: 90923-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR