1-Methyl-1H-indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methylindole-3-carboxaldehyde

A117213-100G A117213-10G A117213-25G A117213-1G A117213-5G
103798-850EA 5 USD
103798-850 103824-722 76797-138 76797-140 76797-142 77311-512 77311-514
1-Methyl-1H-indole-3-carbaldehyde 98%
1-Methyl-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.188 g/mol
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 70…72 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00014570
CAS Number: 19012-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR