1-Methylindole 98%

Supplier: Ambeed
Warning

Synonyms: N-Methylindole, 1-Methyl-1H-indole

A276445-100G A276445-10G A276445-500G A276445-1000G A276445-25G
103809-918EA 314 USD
103809-918 103853-236 76692-946 76692-948 76692-950 77093-090 77093-092
1-Methylindole 98%
1-Methylindole

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 239.3 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005800
CAS Number: 603-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR