1-Methyl-1H-indol-5-ylamine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-n-methylindole, 1-Methyl-1H-indol-5-amine, 5-Amino-1-methyl-1H-indole

A118367-10G A118367-250MG A118367-25G A118367-1G A118367-5G
103799-044EA 26 USD
103799-044 77026-250 77026-252 77026-254 77311-720 77311-722
1-Methyl-1H-indol-5-ylamine 98%
1-Methyl-1H-indol-5-ylamine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Boiling Pt: 328 °C (760 mm Hg)
Melting Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839873
CAS Number: 102308-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR