1-Methyl-1H-indol-4-ol 97%

Supplier: Ambeed
Warning

A127758-250MG A127758-1G
103801-006EA 58 USD
103801-006 77026-918 77313-272
1-Methyl-1H-indol-4-ol 97%
1-Methyl-1H-indol-4-ol

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834885
CAS Number: 7556-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR