1-Methylindazole-5-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-indazole-5-carboxylic acid

A872485-25G A872485-250MG
103841-286EA 75 USD
103841-286 76845-542 77036-754
1-Methylindazole-5-carboxylic acid 96%
1-Methylindazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11901714
CAS Number: 1176754-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR