1-Methyl-1H-indazol-6-amine 95%
Supplier: Ambeed
|
Warning
|
A273060-1G
A273060-250MG
A273060-5G
A273060-100MG
A273060-25G
103813-126EA
29
USD
103813-126
103853-042
77092-838
77166-498
77305-122
77892-531
77892-532
1-Methyl-1H-indazol-6-amine 95%
1-Methyl-1H-indazol-6-amine
Precaución: for Research Use Only
Formula:
C₈H₉N₃ Melting Pt: 172…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00456076 CAS Number: 74728-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...