1-Methyl-1H-indazol-5-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxy-1-methyl-1H-indazole
A159062-100MG
A159062-250MG
A159062-50MG
A159062-5MG
A159062-1G
A159062-5G
103805-222EA
81
USD
103805-222
103843-436
77028-570
77028-572
77028-574
77028-576
77319-308
77319-310
1-Methyl-1H-indazol-5-ol 98%
1-Methyl-1H-indazol-5-ol
Precaución: for Research Use Only
Formula:
C₈H₈N₂O MW: 148.1619 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11869778 CAS Number: 756839-14-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...