1-Methyl-1H-indazol-5-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-methyl-1H-indazole, 1-methyl-1H-indazole-5-amine, 1-METHYL-1H-INDAZOL-5-AMINE

A120142-250MG A120142-25G A120142-1G
103799-892EA 27 USD
103799-892 77161-254 77161-256 77311-996
1-Methyl-1H-indazol-5-amine 97%
1-Methyl-1H-indazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.181 g/mol
Boiling Pt: 321.6 °C (760 mm Hg)
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD03305455
CAS Number: 50593-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR