1-Methylimidazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-imidazole-4-carboxylic acid

A347902-25G A347902-100G A347902-10G A347902-250MG A347902-1G A347902-5G
103825-982EA 10 USD
103825-982 103836-968 76840-042 77239-956 77239-958 77239-960 77306-590 77306-592
1-Methylimidazole-4-carboxylic acid 98%
1-Methylimidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Boiling Pt: 399.1 °C (760 mm Hg)
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD02179560
CAS Number: 41716-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR