1-Methylimidazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-imidazole-4-carbaldehyde

A331805-1G A331805-100MG A331805-250MG
103825-586EA 70 USD
103825-586 103836-552 76839-544 77239-772 77239-774
1-Methylimidazole-4-carbaldehyde 97%
1-Methylimidazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
Boiling Pt: 300.1 °C (760 mm Hg)
Melting Pt: 68…69 °C
Flash Pt: 135.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411957
CAS Number: 17289-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR