1-Methylimidazole-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde

A200864-10G A200864-250MG A200864-1G A200864-25G A200864-5G
103817-246EA 16 USD
103817-246 103847-880 77012-636 77012-638 77085-944 77302-724 77302-726
1-Methylimidazole-2-carbaldehyde 98%
1-Methylimidazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 70…74 °C (1 mm Hg)
Melting Pt: 36…39 °C
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321308
CAS Number: 13750-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR