1-Methyl-2-formylbenzimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazole-2-carbaldehyde

A117208-250MG A117208-5G A117208-1G
103798-846EA 28 USD
103798-846 103824-718 77233-952 77297-520 77311-506
1-Methyl-2-formylbenzimidazole 95%
1-Methyl-2-formylbenzimidazole

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.176 g/mol
Boiling Pt: 332.7 °C (760 mm Hg)
Melting Pt: 114…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142655
CAS Number: 3012-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR