1-Methyl-1H-1,2,4-triazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A452833-100MG A452833-250MG A452833-5G
103827-810EA 333 USD
103827-810 103838-880 77156-140 77156-142 77307-376
1-Methyl-1H-1,2,4-triazole-5-carboxylic acid 97%
1-Methyl-1H-1,2,4-triazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
Boiling Pt: 369.5±25.0 °C (760 mm Hg)
Flash Pt: 177.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100092
CAS Number: 815588-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR