1-Methyl-1H-1,2,4-triazol-3-amine 97%

Supplier: Ambeed
Warning

A158973-100MG A158973-250MG A158973-1G A158973-10G
103805-204EA 34 USD
103805-204 103843-416 76997-688 76997-690 77319-278 77881-240
1-Methyl-1H-1,2,4-triazol-3-amine 97%
1-Methyl-1H-1,2,4-triazol-3-amine

Precaución: for Research Use Only

Formula: C₃H₆N₄
Melting Pt: 81…86 °C
Storage Temperature: Ambient
MDL Number: MFCD02853162
CAS Number: 49607-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR