1-Methyl-1H-1,2,3-triazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A134877-100MG A134877-5G A134877-250MG
103802-062EA 94 USD
103802-062 76811-466 76811-468 77611-208
1-Methyl-1H-1,2,3-triazole-5-carboxylic acid 97%
1-Methyl-1H-1,2,3-triazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD10696347
CAS Number: 716361-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR