1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A136701-10G A136701-25G A136701-5G A136701-100G
103802-332EA 6 USD
103802-332 76782-698 77315-248 77315-250 77666-968
1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
1-Methyl-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798989
CAS Number: 4965-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR