1-Methyl-L-glutamate 98%

Supplier: Ambeed
Warning

Synonyms: H-Glu-OMe, 1-Methyl L-Glutamate, L-Glutamic Acid 1-Methyl Ester

A225098-10G A225098-1G A225098-25G A225098-5G
103819-158EA 14 USD
103819-158 103850-050 76815-018 77303-728 77303-730 77303-732
1-Methyl-L-glutamate 98%
1-Methyl-L-glutamate

Precaución: for Research Use Only

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Boiling Pt: 303.3 °C (760 mm Hg)
Melting Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00171677
CAS Number: 6384-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR