2,3-Dimethylanisole 98%
Supplier: Ambeed
|
Warning
|
A100758-10G
A100758-500G
A100758-5G
A100758-25G
103821-308EA
14
USD
103821-308
77160-176
77160-178
77296-836
77891-748
2,3-Dimethylanisole 98%
2,3-Dimethylanisole
Precaución: for Research Use Only
Formula:
C₉H₁₂O Boiling Pt: 195.7 °C (760 mm Hg) Melting Pt: 29 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008376 CAS Number: 2944-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...