1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Isopropyl-1H-pyrazole-5-boronic acid pinacol ester

A439350-1G A439350-5G A439350-250MG
103827-644EA 17 USD
103827-644 103838-710 76842-464 76842-466 76877-368
1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₂H₂₁BN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD17167306
CAS Number: 1282518-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR