1-Isopropylpyrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1-Isopropyl-1H-pyrazole

A199197-1G A199197-10G A199197-25G A199197-5G
103817-078EA 15 USD
103817-078 103847-702 77085-746 77112-104 77112-106 77302-610
1-Isopropylpyrazole 98%
1-Isopropylpyrazole

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 143 °C (760 mm Hg)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD07468920
CAS Number: 18952-87-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR