1-Iodo-4-phenoxybenzene 98%

Supplier: Ambeed
Warning

A420370-1G A420370-25G A420370-5G
103827-384EA 14 USD
103827-384 103838-432 76842-054 76842-056 76842-058
1-Iodo-4-phenoxybenzene 98%
1-Iodo-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₉IO
MW: 296.1037 g/mol
Boiling Pt: 421.362 °C (760 mm Hg)
Flash Pt: 208.633 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060666
CAS Number: 2974-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR