4-Iodotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-4-methylbenzene

A249740-1000G A249740-25G A249740-100G A249740-10G A249740-1G A249740-500G A249740-5G
103809-772EA 166 USD
103809-772 77091-070 77091-072 77098-650 77098-652 77098-654 77098-656 77098-658
4-Iodotoluene 98%
4-Iodotoluene

Precaución: for Research Use Only

Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 211.5 °C (760 mm Hg)
Melting Pt: 33…36 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR