4-Iodo-3-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-4-methyl-2-nitrobenzene

A161186-100G A161186-5G
103805-464EA 6 USD
103805-464 103843-702 76768-566 77319-668
4-Iodo-3-nitrotoluene 97%
4-Iodo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.03 g/mol
Boiling Pt: 299.3 °C (760 mm Hg)
Melting Pt: 53…56 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00024196
CAS Number: 5326-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR