1-Iodo-4-(trifluoromethoxy)benzene 98%
Supplier: Ambeed
|
Warning
|
A300173-25G
A300173-10G
A300173-5G
A300173-100G
103835-414EA
6
USD
103835-414
76838-106
77032-474
77305-794
77881-830
1-Iodo-4-(trifluoromethoxy)benzene 98%
1-Iodo-4-(trifluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₄F₃IO Boiling Pt: 177…179 °C (760 mm Hg) Flash Pt: 77.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042411 CAS Number: 103962-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...