1-Iodo-4-(4-pentylcyclohexyl)benzene 97%

Supplier: Ambeed
Warning

A650027-1G A650027-5G
103840-394EA 46 USD
103840-394 76844-554 76844-556
1-Iodo-4-(4-pentylcyclohexyl)benzene 97%
1-Iodo-4-(4-pentylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₇H₂₅I
MW: 356.29 g/mol
Boiling Pt: 378.5±21.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00445197
CAS Number: 948553-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR