5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 98%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Di(trifluormethyl)-5-iodbenzene, 1,3-Bis(trifluoromethyl)-5-iodobenzene, 1-Iodo-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)iodobenzene

A176380-100G A176380-10G A176380-500G A176380-25G A176380-5G
103845-456EA 7 USD
103845-456 76769-424 76769-426 76769-428 77082-806 77082-808
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 98%
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene

Precaución: for Research Use Only

Formula: C₈H₃F₆I
MW: 340.01 g/mol
Boiling Pt: 184 °C (760 mm Hg)
Flash Pt: 74.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00040837
CAS Number: 328-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR