1-Iodo-2-phenoxybenzene 98%

Supplier: Ambeed
Warning

A332174-100MG A332174-250MG A332174-5G A332174-1G
103825-598EA 11 USD
103825-598 77239-778 77239-780 77239-782 77306-428
1-Iodo-2-phenoxybenzene 98%
1-Iodo-2-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₉IO
MW: 296.1 g/mol
Boiling Pt: 303.9 °C (760 mm Hg)
Flash Pt: 137.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00767196
CAS Number: 34883-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR