2-Iodotoluene 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A294343-1000G A294343-25G A294343-100G A294343-10G A294343-500G A294343-5G
103809-986EA 313 USD
103809-986 76837-782 76837-784 77114-336 77114-338 77114-340 77114-342
2-Iodotoluene 98% stabilized
2-Iodotoluene

Precaución: for Research Use Only

Formula: C₇H₇I
Boiling Pt: 211.5 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001042
CAS Number: 615-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR